Learn More
Strem, An Ascensus Company 100mg. (4R,4'R,5S,5'S)-2,2'-Cyclobutylidenebis[4,5-dihydro-4,5-diphenyloxazole], 98%.
SDP
Supplier:  Strem, An Ascensus Company 071076100MG
Encompass
100mg. (4R,4'R,5S,5'S)-2,2'-Cyclobutylidenebis[4,5-dihydro-4,5-diphenyloxazole], 98%. . Molecular Weight: 498.6. Molecular Formula: C34H30N2O2. Color/form: off white to light yellow pwdr. Strem# 07-1076. http://www.strem.com/catalog/v/07-1076/
Catalog No. 50-206-5395