Learn More
Strem, An Ascensus Company CAS# 298693-02-4. 25mg. (4S,4'S)-2,2'-Cyclobutylidenebis[4,5-dihydro-4-tert-butyloxazole], 98%.
SDP
Supplier:  Strem, An Ascensus Company 07107925MG
Encompass
CAS# 298693-02-4. 25mg. (4S,4'S)-2,2'-Cyclobutylidenebis[4,5-dihydro-4-tert-butyloxazole], 98%. . Molecular Weight: 306.4. Molecular Formula: C18H30N2O2. Color/form: off white to light yellow pwdr. Strem# 07-1079. http://www.strem.com/catalog/v/07-1079/
Catalog No. 50-206-5397