Learn More
Thomas Scientific Storage Box, 100 x 1.5mL, Black
SDP
Catalog No. 501461062
Encompass
100 place box with hinged lid Polypropylene construction Will not degrade or absorb moisture Withstand freezing (to -80°C) and autoclaving Boxes hold 100 1.5/2.0 mL microtubes. The translucent lid is alphanumerically labeled for easy sample identification. A three-point hinge maintains its integrity during repeated use and a positive catch keeps lid securely fastened. Dimensions: 142 x 142 x 55 mm.
Catalog No. 50-146-1062 Supplier Thomas Scientific Supplier No. 1149X41
100 place box with hinged lid Polypropylene construction Will not degrade or absorb moisture Withstand freezing (to -80°C) and autoclaving Boxes hold 100 1.5/2.0 mL microtubes. The translucent lid is alphanumerically labeled for easy sample identification. A three-point hinge maintains its integrity during repeated use and a positive catch keeps lid securely fastened. Dimensions: 142 x 142 x 55 mm.