Learn More
Biotage BOX OF 96 PHYTIP COL 500UL VOL
SDP
Catalog No. NC1770364
Encompass
Box of 96 PhyTip Columns (500ul volume) containing 400ul of ProPlus affinity resin per column, Tecan MCA.
Catalog No. NC1770364 Supplier Biotage Supplier No. PTT954007
Box of 96 PhyTip Columns (500ul volume) containing 400ul of ProPlus affinity resin per column, Tecan MCA.