Learn More
Biotage BOX OF 96 PHYTIP COLUMNS( 200
SDP
Catalog No. NC1555084
Encompass
Box of 96 PhyTip columns( 200 µL volume) containing 5 µL of Normal Phase I resin per column
Catalog No. NC1555084 Supplier Biotage Supplier No. PTR920509
Box of 96 PhyTip columns( 200 µL volume) containing 5 µL of Normal Phase I resin per column