Learn More
TWD Scientific LLC BioX 100-Well Polycarbonate Freezer Box, 2 inch, Assorted Colors. Case of 20
SDP
Catalog No. 502274268 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 2 inch 100 well polycarbonate freezer box (dimensions: 132x132x53mm) with removable lid. Boxes/lids have numeric grid for identification purposes. Boxes come in assorted colors (red, yellow, green, blue and white). 20/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4268 Supplier TWD Scientific LLC Supplier No. BX9092101
BioX 2 inch 100 well polycarbonate freezer box (dimensions: 132x132x53mm) with removable lid. Boxes/lids have numeric grid for identification purposes. Boxes come in assorted colors (red, yellow, green, blue and white). 20/case, price/case