Learn More
TWD Scientific LLC BioX 81-Well Polycarbonate Freezer Box, 2 inch, Assorted Colors. Case of 20
SDP
Supplier:  TWD Scientific LLC BX9090091
Encompass_Preferred
BioX 2 inch 81 well polycarbonate freezer box (dimensions: 132x132x53mm) with removable lid. Boxes/lids have numeric grid for identification purposes. Boxes come in assorted colors (red, yellow, green, blue and white). 20/case, price/case
Catalog No. 50-227-4267