Learn More
JDM LAB SOLUTIONS LLC BOX SCIENTIFIC STATION ONE
Catalog No. NC1721737
Encompass
The Station One thaws frozen samples and reagents in a fraction of the time using only ambient air. Dual fans intake ambient air and channel it upward through a perforated thawing tray where it circulates over and around samples. The unit includes accessory racks that will fit most industry standard tubes from 1 to 50ml. Equipped with the included accessory racks it can thaw 34 large tubes (15-50ml) or 52 small tubes (1-2ml) in a single cycle.
Catalog No. NC1721737 Supplier JDM LAB SOLUTIONS LLC Supplier No. 13000725
The Station One thaws frozen samples and reagents in a fraction of the time using only ambient air. Dual fans intake ambient air and channel it upward through a perforated thawing tray where it circulates over and around samples. The unit includes accessory racks that will fit most industry standard tubes from 1 to 50ml. Equipped with the included accessory racks it can thaw 34 large tubes (15-50ml) or 52 small tubes (1-2ml) in a single cycle.