Learn More
Electron Microscopy Sciences Cryogenic Grid Box with Lid, Square for Gatan CT3500, White
SDP
Catalog No. 501929316
Encompass_Preferred
These specimen grid boxes are tools for storing or transferring cryogenic TEM specimen grids. Features four diamond shaped slots. Non-static clear covershield held in place with stainless steel screw, which is tapped in the center of the box. Box fits the FEI Vitrobot™, Gatan 626™, Gatan 3500™.
Color: White
Catalog No. 50-192-9316 Supplier Electron Microscopy Sciences Supplier No. 7116660
These specimen grid boxes are tools for storing or transferring cryogenic TEM specimen grids. Features four diamond shaped slots. Non-static clear covershield held in place with stainless steel screw, which is tapped in the center of the box. Box fits the FEI Vitrobot™, Gatan 626™, Gatan 3500™.
Color: White