Learn More
Selleck Chemical LLC Box5 P1216-100MG
SDP
Catalog No. 5000041550
Encompass_Preferred
Box5 a Wnt5a-derived N-butyloxycarbonyl hexapeptide (t-Boc-Met-Asp-Gly-Cys-Glu-Leu) acts as an Wnt5a antagonist inhibits the basal migration and invasion of Wnt5a-expressing HTB63 melanoma cells antagonizes the effects of Wnt5a on melanoma cell migration and invasion by directly inhibiting Wnt5a-induced PKC and Ca(2 ) signaling
Catalog No. 50-000-41550 Supplier Selleck Chemical LLC Supplier No. P1216100MG
Box5 a Wnt5a-derived N-butyloxycarbonyl hexapeptide (t-Boc-Met-Asp-Gly-Cys-Glu-Leu) acts as an Wnt5a antagonist inhibits the basal migration and invasion of Wnt5a-expressing HTB63 melanoma cells antagonizes the effects of Wnt5a on melanoma cell migration and invasion by directly inhibiting Wnt5a-induced PKC and Ca(2 ) signaling