Learn More
Medchemexpress LLC Box5 (TFA) | 00-00-0 | 99.2% | 880.9 Da | C32H51F3N6O15S2.xC2HF3O2 | 5 MG
SDP
Catalog No. 5000264674
Encompass_Preferred
Box5 TFA is the trifluoroacetate salt form of a peptide antagonist of Wnt5a used in vitro to study Wnt5a-mediated signaling, calcium release, and cell migration. Supplied in small milligram vials, the material is provided with documentation to support research use.
  • Trifluoroacetate salt form for improved solubility and handling.
  • Potent antagonist of Wnt5a signaling for pathway studies.
  • Inhibits Wnt5a-mediated calcium release and cell migration.
  • High purity approximately 99.2% for consistent results.
  • Supplied in small milligram sizes suitable for in vitro experiments.
  • COA and SDS available for quality and safety verification.
Catalog No. 50-002-64674 Supplier Medchemexpress LLC Supplier No. HY123071A5MG
Box5 TFA is the trifluoroacetate salt form of a peptide antagonist of Wnt5a used in vitro to study Wnt5a-mediated signaling, calcium release, and cell migration. Supplied in small milligram vials, the material is provided with documentation to support research use.
  • Trifluoroacetate salt form for improved solubility and handling.
  • Potent antagonist of Wnt5a signaling for pathway studies.
  • Inhibits Wnt5a-mediated calcium release and cell migration.
  • High purity approximately 99.2% for consistent results.
  • Supplied in small milligram sizes suitable for in vitro experiments.
  • COA and SDS available for quality and safety verification.