Learn More
Strem, An Ascensus Company CAS# 1358991-79-3. 100mg. (1Z,3Z)-1,3-Bis[[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl]methylene]-2,3-dihydro-5,6-dimethyl-1H-isoindole, 95%.
SDP
Catalog No. 502065391
Encompass
CAS# 1358991-79-3. 100mg. (1Z,3Z)-1,3-Bis[[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl]methylene]-2,3-dihydro-5,6-dimethyl-1H-isoindole, 95%. . Molecular Weight: 461.6. Molecular Formula: C30H27N3O2. Color/form: light yellow to yellow pwdr. Strem# 07-1070. http://www.strem.com/catalog/v/07-1070/
Catalog No. 50-206-5391 Supplier Strem, An Ascensus Company Supplier No. 071070100MG
Add to Cart
Add to Cart
CAS# 1358991-79-3. 100mg. (1Z,3Z)-1,3-Bis[[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl]methylene]-2,3-dihydro-5,6-dimethyl-1H-isoindole, 95%. . Molecular Weight: 461.6. Molecular Formula: C30H27N3O2. Color/form: light yellow to yellow pwdr. Strem# 07-1070. http://www.strem.com/catalog/v/07-1070/