Learn More
Med Vet International Miltex Vantage Bozeman Dressing Forceps, 10.5in., Double Curved, Serrated Tips
Catalog No. 502831950
Encompass_Preferred
The Miltex Vantage V97-622 Bozeman dressing forceps are made from high quality Japanese stainless steel and measure 10.5" in length. They have a finger ring handle with a ratchet lock and double-curved, serrated tips.
Catalog No. 50-283-1950 Supplier Med Vet International Supplier No. MILV97622
The Miltex Vantage V97-622 Bozeman dressing forceps are made from high quality Japanese stainless steel and measure 10.5" in length. They have a finger ring handle with a ratchet lock and double-curved, serrated tips.