Learn More
Sklar Instruments Forceps, Bozeman Forceps, Sklar 10-1/4 in., Stainless Steel, EA.
SDP
Catalog No. 501121752 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Bozeman Forceps, Sklar, 10-1/4 in., Stainless Steel, EA. Sklar Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures. They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. This product features an in.S in. curve (slightly curved shanks with jaws slightly curved opposite direction) with partial horizontal serrations and a length of 10-1/4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-1752 Supplier Sklar Instruments Supplier No. 801410
Forceps, Bozeman Forceps, Sklar, 10-1/4 in., Stainless Steel, EA. Sklar Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures. They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. This product features an in.S in. curve (slightly curved shanks with jaws slightly curved opposite direction) with partial horizontal serrations and a length of 10-1/4 inches. Lifetime warranty against all manufacturer defects.