Learn More
Sklar Instruments Econo Bozeman Forceps, Sklar, 10 in., Curved, Serrated, Stainless Steel. PK/12.
SDP
Catalog No. 501437628 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Bozeman Forceps, Sklar, 10 in., Curved, Serrated, Stainless Steel. Econo Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures. They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product features an in.S in. curve (slightly curved shanks with jaws slightly curved opposite direction) with partial horizontal serrations and a length of 10 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7628 Supplier Sklar Instruments Supplier No. 21374
Econo Bozeman Forceps, Sklar, 10 in., Curved, Serrated, Stainless Steel. Econo Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures. They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product features an in.S in. curve (slightly curved shanks with jaws slightly curved opposite direction) with partial horizontal serrations and a length of 10 inches. They are sold in boxes of 12.