Learn More
Medchemexpress LLC Bozitinib | 1440964-89-5 | MFCD00059343 | 98.9% | 424.38 g/mol | C20H15F3N8 | 25MG
SDP
Catalog No. 5000237890
Encompass_Preferred
Bozitinib is a selective, ATP-competitive small-molecule inhibitor of the c-Met (HGFR) tyrosine kinase intended for laboratory research. It demonstrates blood-brain barrier permeability and is used in preclinical studies to probe c-Met signaling and downstream pathways. Supplied as a solid for research use only, handle and store according to the supplier datasheet.
Catalog No. 50-002-37890 Supplier Medchemexpress LLC Supplier No. HY12501725MG
Bozitinib is a selective, ATP-competitive small-molecule inhibitor of the c-Met (HGFR) tyrosine kinase intended for laboratory research. It demonstrates blood-brain barrier permeability and is used in preclinical studies to probe c-Met signaling and downstream pathways. Supplied as a solid for research use only, handle and store according to the supplier datasheet.