Learn More
TARGETMOL CHEMICALS INC BOZITINIB 5MG
SDP
Catalog No. 502581706
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bozitinib (PLB-1001) (PLB-1001) is a highly selective inhibitor of the c-MET kinase with blood-brain barrier permeability. Bozitinib binds to the conventional ATP-binding pocket of the tyrosine kinase superfamily. purity: 99%
Catalog No. 50-258-1706 Supplier TARGETMOL CHEMICALS INC Supplier No. T105855MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bozitinib (PLB-1001) (PLB-1001) is a highly selective inhibitor of the c-MET kinase with blood-brain barrier permeability. Bozitinib binds to the conventional ATP-binding pocket of the tyrosine kinase superfamily. purity: 99%