Learn More
Medchemexpress LLC Bozitinib (PLB-1001) | 1440964-89-5 | 98.9% | 424.38 g/mol | C20H15F3N8 | 100MG
SDP
Catalog No. 5000235892
Encompass_Preferred
Bozitinib (PLB-1001) is a selective, ATP-competitive small-molecule inhibitor of the c-MET (HGFR) tyrosine kinase with demonstrated blood-brain barrier permeability. Supplied for research use in multiple formats and sizes, it is intended for in vitro and preclinical studies of MET-driven oncogenic signaling and kinase pharmacology.
  • Selective c-MET kinase inhibition for MET-driven research.
  • ATP-competitive mechanism of action.
  • Demonstrated blood-brain barrier permeability.
  • Available as solid or DMSO solution formats for assay flexibility.
  • High purity suitable for biochemical and cellular assays.
  • Documented molecular formula and molecular weight for analytical reference.
Catalog No. 50-002-35892 Supplier Medchemexpress LLC Supplier No. HY125017100MG
Bozitinib (PLB-1001) is a selective, ATP-competitive small-molecule inhibitor of the c-MET (HGFR) tyrosine kinase with demonstrated blood-brain barrier permeability. Supplied for research use in multiple formats and sizes, it is intended for in vitro and preclinical studies of MET-driven oncogenic signaling and kinase pharmacology.
  • Selective c-MET kinase inhibition for MET-driven research.
  • ATP-competitive mechanism of action.
  • Demonstrated blood-brain barrier permeability.
  • Available as solid or DMSO solution formats for assay flexibility.
  • High purity suitable for biochemical and cellular assays.
  • Documented molecular formula and molecular weight for analytical reference.