Learn More
Medchemexpress LLC BP 897 hydrochloride 100mg | 314776-92-6 | 100MG
SDP
Catalog No. 5000240173
Encompass_Preferred
BP 897 hydrochloride is a potent and partial dopamine D3 receptor agonist and a weak D2 receptor antagonist BP 897 hydrochloride displays a high affinity at the dopamine D3 receptor (Ki 0 92 nM) and a 70 times lower affinity at the D2 receptor (Ki 61 nM)[1
Catalog No. 50-002-40173 Supplier Medchemexpress LLC Supplier No. HY106660100MG
BP 897 hydrochloride is a potent and partial dopamine D3 receptor agonist and a weak D2 receptor antagonist BP 897 hydrochloride displays a high affinity at the dopamine D3 receptor (Ki 0 92 nM) and a 70 times lower affinity at the D2 receptor (Ki 61 nM)[1