Learn More
Sino Biological BP Cloning Enzyme Kit 100 Tests
Catalog No. 5000696228
Encompass_Preferred
BP Cloning Enzyme Kit is a proprietary enzyme and buffer formulation containing the bacteriophage lambda recombination protein Integrase (Int), the E. coli-encoded protein Integration Host Factor (IHF), and reaction buffer provided in a single mix for convenient reaction set up.
Catalog No. 50-006-96228 Supplier Sino Biological Supplier No. SBBP01100
BP Cloning Enzyme Kit is a proprietary enzyme and buffer formulation containing the bacteriophage lambda recombination protein Integrase (Int), the E. coli-encoded protein Integration Host Factor (IHF), and reaction buffer provided in a single mix for convenient reaction set up.