Learn More
Med Vet International Omron 10 Series Home Automatic Digital Blood Pressure Monitor, Wide Range, Cloth Fabric, 23 - 43 cm, Desk Model, BP7450, Each
Catalog No. 503045570
Encompass_Preferred
Adult large cuff for arm - 22 to 43 cm Black cloth fabric This premium blood pressure monitor stores 200 blood pressure readings for two users (100 per user) With the monitor, you get the OMRON pre-formed ComFit cuff Unit operates on 4 AA batteries (included) and AC Adapter (included)
Catalog No. 50-304-5570 Supplier Med Vet International Supplier No. 1150421EA
Adult large cuff for arm - 22 to 43 cm Black cloth fabric This premium blood pressure monitor stores 200 blood pressure readings for two users (100 per user) With the monitor, you get the OMRON pre-formed ComFit cuff Unit operates on 4 AA batteries (included) and AC Adapter (included)