Learn More
Med Vet International Omron 5 Series Home Automatic Digital Blood Pressure Monitor, Wide Range, Nylon, 23 - 43 cm, Desk Model, BP7250, Each
Catalog No. 503045559
Encompass_Preferred
Advance technology targets several points to obtain additional data for reliable, accurate readings Breathing and body movements don't affect the Omron 5 Series blood pressure monitor during measurements Monitor includes a D-ring style cuff that can accommodate users with an arm circumference from 9 inches to 17 inches Wireless Bluetooth lets you connect to an IOS or Android device using the Omron Connect app The Omron wireless blood pressure monitor alerts you if your heartbeat is irregular or at risk for hypertension
Catalog No. 50-304-5559 Supplier Med Vet International Supplier No. 1150426EA
Advance technology targets several points to obtain additional data for reliable, accurate readings Breathing and body movements don't affect the Omron 5 Series blood pressure monitor during measurements Monitor includes a D-ring style cuff that can accommodate users with an arm circumference from 9 inches to 17 inches Wireless Bluetooth lets you connect to an IOS or Android device using the Omron Connect app The Omron wireless blood pressure monitor alerts you if your heartbeat is irregular or at risk for hypertension