Learn More
Med Vet International Automatic Digital Blood Pressure Monitor ProBP 2000™ Adult Cuff Nylon Cuff Desk Model, 2000-A, Each
Catalog No. 503045557
Encompass_Preferred
Combined two of the innovative technologies; SureBP and FlexiPort which supports accurate blood pressure readings SureBP technology, is a blood pressure averaging algorithm that allows the device to capture blood pressure readings on cuff inflation in a comfortable way for patients Blood pressure readings are captured on inflation in about 20 seconds Meets AAMI standards for blood pressure accuracy Compatible with Welch Allyn FlexiPort Reusable Blood Pressure Cuffs, from child size 9 up to thigh size 13 (15 - 55 cm range) One-button touch triggers the device to start capturing on cuff inflation Single-point connection helps remove the hassle of traditional cuff tubes and connectors, to better support a busy workflow
Catalog No. 50-304-5557 Supplier Med Vet International Supplier No. 1166240EA
Combined two of the innovative technologies; SureBP and FlexiPort which supports accurate blood pressure readings SureBP technology, is a blood pressure averaging algorithm that allows the device to capture blood pressure readings on cuff inflation in a comfortable way for patients Blood pressure readings are captured on inflation in about 20 seconds Meets AAMI standards for blood pressure accuracy Compatible with Welch Allyn FlexiPort Reusable Blood Pressure Cuffs, from child size 9 up to thigh size 13 (15 - 55 cm range) One-button touch triggers the device to start capturing on cuff inflation Single-point connection helps remove the hassle of traditional cuff tubes and connectors, to better support a busy workflow