Learn More
Research Products International Corp BPA, 1 G
SDP
Supplier:  Research Products International Corp B300101.0
Encompass_Preferred
Highly mobile synthetic cytokinin. Typically applied as foliar spray. Increases branching and growth.
Catalog No. 50-197-7059