Learn More
Research Products International Corp BPA [n-Benzyl-9-(2-tetrahydropyranyl)-adenine], 100 Milligrams
SDP
Supplier:  Research Products International Corp B300100.1
Encompass_Preferred
Highly mobile synthetic cytokinin. Typically applied as foliar spray. Increases branching and growth.
Catalog No. 50-213-159