Learn More
Waters Corp Column Kit, for Bisphenol A Analysis, ASTM D7574-09 Compliant, ACQUITY UPLC System Optimized
Supplier:  Waters Corp 176001955
Encompass_Preferred
This solution for the analysis of Bisphenol A in drinking water is fully compliant with ASTM Method D7574-09. This kit is optimized for use with the UPLC system.
  • Optimized for use with the UPLC system.
  • Compliant with ASTM Method D7574-09 for Bisphenol A analysis in drinking water.
  • Includes a BEH C18, 1.7 μm, 2.1 × 50 mm column.
  • Includes an isolator column.
Catalog No. 50-791-073