Learn More
Waters Corp BPA Method Kit 186004932
Supplier:  Waters Corp 186004932
Encompass_Preferred
Waters ACQUITY UPLC solution for the analysis of Bisphenol A in Drinking Water is fully compliant with ASTM Method D7574-09. Optimized for use with the ACQUITY system, this kit contains: ACQUITY BEH C18, 1.7um, 2.1 x 50mm Column; ACQUITY Isolator Column; Oasis HLB 5cc (200mg) LP Glass Cartridges and LCMS Certified Vials. Included in ASTM Method D7574-09, these products have been selected to ensure to highest performance available.
Catalog No. 50-791-245