Learn More
Alkali Scientific BPA [n Benzyl 9 (2 tetrahydropyranyl) adenine], 1 G
SDP
Catalog No. 5000660307
Encompass_Preferred
Benzyl Adenine (BPA) is a powerful plant growth regulator used in laboratory settings to stimulate shoot proliferation and enhance growth in plant tissues. It is widely used as a cytokinin to promote cell division and extend the lifespan of plant cells. BPA is especially valuable in plant tissue culture, helping researchers to propagate plants and regenerate new growth more effectively. This powder form of BPA is commonly used in agricultural and horticultural research, particularly for enhancing tissue regeneration, improving clonal propagation, and optimizing crop yields. BPA is a vital reagent for plant biotechnologists and horticulturists focused on plant development and genetic research.
Catalog No. 50-006-60307 Supplier Alkali Scientific Supplier No. BPA002CP
Benzyl Adenine (BPA) is a powerful plant growth regulator used in laboratory settings to stimulate shoot proliferation and enhance growth in plant tissues. It is widely used as a cytokinin to promote cell division and extend the lifespan of plant cells. BPA is especially valuable in plant tissue culture, helping researchers to propagate plants and regenerate new growth more effectively. This powder form of BPA is commonly used in agricultural and horticultural research, particularly for enhancing tissue regeneration, improving clonal propagation, and optimizing crop yields. BPA is a vital reagent for plant biotechnologists and horticulturists focused on plant development and genetic research.