Learn More
Alkali Scientific BPA [n Benzyl 9 (2 tetrahydropyranyl) adenine], 100 Mg
SDP
Catalog No. 5000661504
Encompass_Preferred
Benzyl Adenine (BPA) is a plant growth regulator in powder form, used extensively in laboratories to promote cell division and enhance plant growth. This cytokinin is known to stimulate shoot formation and can delay senescence in plant tissues. BPA is typically used in tissue culture and plant regeneration studies to enhance shoot proliferation and improve cloning efficiency in various plant species. By promoting cell division and modulating growth, BPA helps researchers and horticulturists achieve higher success rates in propagation and genetic improvement programs. It is widely applied in agricultural research, particularly in crop improvement and plant biotechnology.
Catalog No. 50-006-61504 Supplier Alkali Scientific Supplier No. BPA001CP
Benzyl Adenine (BPA) is a plant growth regulator in powder form, used extensively in laboratories to promote cell division and enhance plant growth. This cytokinin is known to stimulate shoot formation and can delay senescence in plant tissues. BPA is typically used in tissue culture and plant regeneration studies to enhance shoot proliferation and improve cloning efficiency in various plant species. By promoting cell division and modulating growth, BPA helps researchers and horticulturists achieve higher success rates in propagation and genetic improvement programs. It is widely applied in agricultural research, particularly in crop improvement and plant biotechnology.