Learn More
Medchemexpress LLC BPAM344 | 1204572-55-3 | 99.7% | 50 MG
SDP
Catalog No. 5000339392
Encompass_Preferred
BPAM344 is a positive allosteric modulator (PAM) of kainate receptor (KAR) subunits GluK1b, GluK2a, and GluK3a. It is intended for research use only and not for sale to patients.
  • Potentiates glutamate-evoked currents of GluK2a 21-fold at 200 μM
  • Significantly decreases desensitization kinetics
  • Potentiates peak current amplitude of KAR subunits GluK3a, GluK2a, and GluK1b
  • Potentiates AMPA receptor subunit GluA1i
  • Exhibits EC50 of 0.9 μM for positive allosteric modulation of AMPA receptor in primary neuronal culture
Catalog No. 50-003-39392 Supplier Medchemexpress LLC Supplier No. HY12908650MG
BPAM344 is a positive allosteric modulator (PAM) of kainate receptor (KAR) subunits GluK1b, GluK2a, and GluK3a. It is intended for research use only and not for sale to patients.
  • Potentiates glutamate-evoked currents of GluK2a 21-fold at 200 μM
  • Significantly decreases desensitization kinetics
  • Potentiates peak current amplitude of KAR subunits GluK3a, GluK2a, and GluK1b
  • Potentiates AMPA receptor subunit GluA1i
  • Exhibits EC50 of 0.9 μM for positive allosteric modulation of AMPA receptor in primary neuronal culture