Learn More
Medchemexpress LLC HY-W014815 500mg Medchemexpress, BPBA CAS:223463-14-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW014815500MG
Encompass_Preferred
Medchemexpress, HY-W014815 500mg BPBA CAS:223463-14-7 BPBA (2-Bromopyridine-5-boronic acid) is a new labeling reagent to derivatize brassinosteroids (BRs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3367