Learn More
Being Scientific BPC-13A - 13L, –20℃ to 150℃, 5L /min flow rate
Supplier:  Being Scientific BCH15013AU
Encompass_Preferred
The BEING BPC Series cooling/heating circulators provideconsistent, precise, and stable laboratory temperature controlfor cooling or heating through constantly circulating water. It canbe used for supplying auxiliary heating for water baths. They arewidely used in various applications like dissolution testing, andsample and reagent heating in colleges, industrial and miningenterprises, and scientifi c research departments.
Catalog No. 50-002-17557