Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bpc 157 acetate | 1628202-19-6 | 99.8% | 1419.54 (free acid) | C62H98N16O22 · x C2H4O2 | 100 MG
SDP

Catalog No. 5000243570
Encompass_Preferred

BPC 157 acetate is the acetate salt of a 15-amino-acid gastric peptide supplied as a research reagent for in vitro and biochemical studies. It is commonly used in investigations of wound healing, gastrointestinal protection, and neuroprotection due to its reported biological activity and high chemical purity.

  • High purity 99.8%.
  • Peptide sequence: GEPPPGKPADDAGLV.
  • Molecular weight 1419.54 (free acid).
  • Supplied as a lyophilized acetate salt suitable for laboratory use.
  • Available in small research quantities including 100 MG packs.

Catalog No. 50-002-43570 Supplier Medchemexpress LLC Supplier No. HY105174A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BPC 157 acetate is the acetate salt of a 15-amino-acid gastric peptide supplied as a research reagent for in vitro and biochemical studies. It is commonly used in investigations of wound healing, gastrointestinal protection, and neuroprotection due to its reported biological activity and high chemical purity.

  • High purity 99.8%.
  • Peptide sequence: GEPPPGKPADDAGLV.
  • Molecular weight 1419.54 (free acid).
  • Supplied as a lyophilized acetate salt suitable for laboratory use.
  • Available in small research quantities including 100 MG packs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.