Learn More
Medchemexpress LLC Bpc 157 acetate | 1628202-19-6 | 99.8% | 1419.54 (free acid) | C62H98N16O22.xC2H4O2 | 10 MG
SDP
Catalog No. 5000243206
Encompass_Preferred
Bpc 157 acetate is the acetate salt form of the 15-amino-acid gastric pentadecapeptide BPC-157 supplied for research use. It is provided as a lyophilized peptide for use in in vitro and in vivo experimental studies, with reported wound healing and neuroprotective activities.
  • Lyophilized peptide suitable for laboratory use.
  • High purity: 99.8%.
  • Molecular weight: 1419.54 (free acid).
  • Available in small research sizes, e.g., 10 mg.
  • RP-HPLC documentation available for purity verification.
Catalog No. 50-002-43206 Supplier Medchemexpress LLC Supplier No. HY105174A10MG
Bpc 157 acetate is the acetate salt form of the 15-amino-acid gastric pentadecapeptide BPC-157 supplied for research use. It is provided as a lyophilized peptide for use in in vitro and in vivo experimental studies, with reported wound healing and neuroprotective activities.
  • Lyophilized peptide suitable for laboratory use.
  • High purity: 99.8%.
  • Molecular weight: 1419.54 (free acid).
  • Available in small research sizes, e.g., 10 mg.
  • RP-HPLC documentation available for purity verification.