Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BPC 157 acetate | 1628202-19-6 | 99.8% | 1419.54 g/mol (free acid) | C62H98N16O22 · x C2H4O2 | 25 MG
SDP

Catalog No. 5000243890
Encompass_Preferred

BPC 157 acetate is the acetate salt of a 15-amino-acid gastric peptide fragment used for research. Supplied as a white to off-white powder, it is reported to promote wound healing, provide neuroprotective activity, and help maintain gastrointestinal mucosal integrity. The material is supplied at high purity and requires sealed, cold storage for long-term stability.

  • High purity (~99.8%).
  • Acetate salt, white to off-white solid.
  • Molecular weight 1419.54 g/mol (free acid).
  • Peptide sequence: Gly-Glu-Pro-Pro-Pro-Gly-Lys-Pro-Ala-Asp-Asp-Ala-Gly-Leu-Val.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-43890 Supplier Medchemexpress LLC Supplier No. HY105174A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BPC 157 acetate is the acetate salt of a 15-amino-acid gastric peptide fragment used for research. Supplied as a white to off-white powder, it is reported to promote wound healing, provide neuroprotective activity, and help maintain gastrointestinal mucosal integrity. The material is supplied at high purity and requires sealed, cold storage for long-term stability.

  • High purity (~99.8%).
  • Acetate salt, white to off-white solid.
  • Molecular weight 1419.54 g/mol (free acid).
  • Peptide sequence: Gly-Glu-Pro-Pro-Pro-Gly-Lys-Pro-Ala-Asp-Asp-Ala-Gly-Leu-Val.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.