Learn More
Cayman Chemical BPC 157acetate 25mg
SDP
Catalog No. 502989683
Encompass_Preferred
A BPC pentadecapeptide fragment with diverse diverse biological activities; increases cell migration of, and F-actin formation in, primary rat tendon fibroblasts at 2 UG/ml; reduces paw swelling, bone erosion, and inflamed joint mononuclear cell infiltration in a rat model of CFA-induced rheumatoid arthritis at 0.01 and 10 UG/kg; decreases the lesion size of indomethacin-, aspirin-, or diclofenac-induced gastric ulcers in rats at 0.01 and 10 UG/kg; decreases the duration of catalepsy and reduces tremor severity in an MPTP-induced mouse model of Parkinson's disease at 0.01 and 10 UG/kg
Catalog No. 50-298-9683 Supplier Cayman Chemical Supplier No. 3098925 MG
A BPC pentadecapeptide fragment with diverse diverse biological activities; increases cell migration of, and F-actin formation in, primary rat tendon fibroblasts at 2 UG/ml; reduces paw swelling, bone erosion, and inflamed joint mononuclear cell infiltration in a rat model of CFA-induced rheumatoid arthritis at 0.01 and 10 UG/kg; decreases the lesion size of indomethacin-, aspirin-, or diclofenac-induced gastric ulcers in rats at 0.01 and 10 UG/kg; decreases the duration of catalepsy and reduces tremor severity in an MPTP-induced mouse model of Parkinson's disease at 0.01 and 10 UG/kg