Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BPC ReadyMate™ to Female Opta™ Tubing Set

Catalog No. bp0060a
Change view
Click to view available options
Description:
BPC ReadyMate to Female Opta Tubing Set
Length (Metric):
1 m
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Length (Metric) Quantity
BP0060A BPC ReadyMate to Female Opta Tubing Set 1 m 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP0060A Supplier Thermo Scientific™ Supplier No. BP0060A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Minimize prep time and maximize production for media fill trials with ready-to-use media in BPCs. Multiple connections give greater flexibility.

Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to Female Opta Tubing Set.

  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labor costs

Multiple plug-and-play connection options, such as BPC ReadyMate to Female Opta, are available for greater flexibility.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Opta™ is a trademark or registered trademark of the Sartorius Stedim Biotech.

ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.

TRUSTED_SUSTAINABILITY

Specifications

Color Multi
Description BPC ReadyMate to Female Opta Tubing Set
Format Bioprocess Container
Product Type Microbiology Media
Includes Tubing and connectors
Quantity 1/Pk.
Length (Metric) 1 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.