Learn More
Enzo Life Sciences BPDEtide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1120001
Encompass_Preferred
Sequence derived from bovine lung cGMP specific phosphodiesterase (cG-BPDE). Specific substrate for PKG, Km=68 µM; Km for PKA=320 µM, with no phosphorylation detected for CaMKII or PKC. Alternative name: cGMP-dependent protein kinase (PKG) substrate. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1232