Learn More
TARGETMOL CHEMICALS INC BPHA 5MG
SDP
Catalog No. 502580169
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BPHA (MMP-2/MMP-9 Inhibitor II) is a potent selective and orally active inhibitor of MMP-2 MMP-9 and MMP-14 with IC50s of 12 nM 16 nM and 17 nM respectively.BPHA does not inhibit MMP-1 -3 and -7 (IC50s of 974 >1000 and 795 nM respectively).BPHA has anti-angiogenic and BPHA has anti-angiogenic and anti-tumor activities. purity: 99%
Catalog No. 50-258-0169 Supplier TARGETMOL CHEMICALS INC Supplier No. T367125MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BPHA (MMP-2/MMP-9 Inhibitor II) is a potent selective and orally active inhibitor of MMP-2 MMP-9 and MMP-14 with IC50s of 12 nM 16 nM and 17 nM respectively.BPHA does not inhibit MMP-1 -3 and -7 (IC50s of 974 >1000 and 795 nM respectively).BPHA has anti-angiogenic and BPHA has anti-angiogenic and anti-tumor activities. purity: 99%