Learn More
Aviva Systems Biology Corp BPHL Antibody - N-terminal region (ARP61857_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61857_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BPHL. It was validated on Western Blot. . Alternate names: BPH-RP, MCNAA, MGC125930, MGC41865, VACVASEApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-487