Learn More
Athens Research And Technology Inc BPI HU NEUTROPHIL 100UG
Supplier:  Athens Research And Technology Inc 1614021609100UG
Encompass_Preferred
BPI, Human Neutrophil (CAP57): 100 UG; Frozen in 50 mM Tris buffer, pH 8.0, with 800 mM NaCl; MW 55,000; EC= 1.0; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from whole blood in the Athens Research & Tech lab.
Catalog No. 50-489-672