Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bpin-Cyclohexene-COOEt | 1049004-32-1 | 280.17 | 1 G
SDP

Catalog No. 5000448576
Encompass_Preferred

Bpin-Cyclohexene-COOEt is a PROTAC linker belonging to the PEG class. This compound can be utilized in the synthesis of PROTAC molecules.

  • PROTAC linker
  • Belongs to the PEG class
  • Suitable for synthesizing PROTAC molecules
  • Purity of ≥97.0% by NMR
  • Appears as a colorless to light yellow liquid
  • Recommended storage at -20°C for 3 years (unstable in solutions, fresh preparation recommended)

Catalog No. 50-004-48576 Supplier Medchemexpress LLC Supplier No. HYW0066351G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bpin-Cyclohexene-COOEt is a PROTAC linker belonging to the PEG class. This compound can be utilized in the synthesis of PROTAC molecules.

  • PROTAC linker
  • Belongs to the PEG class
  • Suitable for synthesizing PROTAC molecules
  • Purity of ≥97.0% by NMR
  • Appears as a colorless to light yellow liquid
  • Recommended storage at -20°C for 3 years (unstable in solutions, fresh preparation recommended)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.