Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bpin-Cyclohexene-COOEt | 1049004-32-1 | 280.17 | 25 G
SDP

Catalog No. 5000449869
Encompass_Preferred

Bpin-Cyclohexene-COOEt is a PROTAC linker from the PEG class, primarily used in the synthesis of PROTAC molecules. This chemical appears as a colorless to light yellow liquid, suitable for laboratory research and the manufacture of substances. It is important to note that this product is intended for research purposes only and has not been validated for medical applications.

  • PROTAC linker for synthesizing PROTAC molecules
  • Clear to light yellow liquid appearance
  • Purity greater than or equal to 97.0%
  • Molecular weight of 280.17
  • Store pure form at -20°C for up to 3 years
  • Unstable in solutions; use freshly prepared solutions
  • Stable under recommended storage conditions
  • Intended for research use only
  • Not validated for medical applications

Catalog No. 50-004-49869 Supplier Medchemexpress LLC Supplier No. 25GHYW006635
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bpin-Cyclohexene-COOEt is a PROTAC linker from the PEG class, primarily used in the synthesis of PROTAC molecules. This chemical appears as a colorless to light yellow liquid, suitable for laboratory research and the manufacture of substances. It is important to note that this product is intended for research purposes only and has not been validated for medical applications.

  • PROTAC linker for synthesizing PROTAC molecules
  • Clear to light yellow liquid appearance
  • Purity greater than or equal to 97.0%
  • Molecular weight of 280.17
  • Store pure form at -20°C for up to 3 years
  • Unstable in solutions; use freshly prepared solutions
  • Stable under recommended storage conditions
  • Intended for research use only
  • Not validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.