Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bpin-Cyclohexene-COOEt, CAS [[1049004-32-1]] | 1049004-32-1 | 280.17 | 10 G
SDP

Catalog No. 5000446178
Encompass_Preferred

Bpin-Cyclohexene-COOEt (Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-cyclohexene-1-carboxylate) is a PROTAC linker belonging to the PEG class. It can be used to synthesize PROTAC molecules.

  • PROTAC linker
  • Belongs to the PEG class
  • Used to synthesize PROTAC molecules
  • Molecular weight: 280.17
  • Purity: 97.78
  • Reference compound

Catalog No. 50-004-46178 Supplier Medchemexpress LLC Supplier No. HYW00663510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bpin-Cyclohexene-COOEt (Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-cyclohexene-1-carboxylate) is a PROTAC linker belonging to the PEG class. It can be used to synthesize PROTAC molecules.

  • PROTAC linker
  • Belongs to the PEG class
  • Used to synthesize PROTAC molecules
  • Molecular weight: 280.17
  • Purity: 97.78
  • Reference compound

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.