Learn More
Medchemexpress LLC BPK-25 | 2305052-86-0 | 99.9% | C21H17ClN4O2 | 50 MG
SDP
Catalog No. 5000354902
Encompass_Preferred
BPK-25 is an active acrylamide that promotes the degradation of nucleosome remodeling and deacetylation (NuRD) complex proteins through a post-translational mechanism involving covalent protein engagement. It also inhibits TMEM173 activation by the cyclic dinucleotide ligand cGAMP.
  • Promotes degradation of NuRD complex proteins.
  • Inhibits TMEM173 activation.
  • Suppresses activation of NF-κB.
  • Blocks nuclear factor of activated T-cells (NFAT) activation.
  • Reduces NFATc2 expression in T cells.
  • Striking and selective reduction of several proteins in the NuRD complex in a concentration- and time-dependent manner.
Catalog No. 50-003-54902 Supplier Medchemexpress LLC Supplier No. HY14155050MG
BPK-25 is an active acrylamide that promotes the degradation of nucleosome remodeling and deacetylation (NuRD) complex proteins through a post-translational mechanism involving covalent protein engagement. It also inhibits TMEM173 activation by the cyclic dinucleotide ligand cGAMP.
  • Promotes degradation of NuRD complex proteins.
  • Inhibits TMEM173 activation.
  • Suppresses activation of NF-κB.
  • Blocks nuclear factor of activated T-cells (NFAT) activation.
  • Reduces NFATc2 expression in T cells.
  • Striking and selective reduction of several proteins in the NuRD complex in a concentration- and time-dependent manner.