Learn More
New England Biolabs, Inc. BpmI – 100 units
SDP
Catalog No. 50812253
Encompass
A restriction endonuclease that recognizes the sequence CTGGAGNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-253 Supplier New England Biolabs, Inc. Supplier No. R0565S
A restriction endonuclease that recognizes the sequence CTGGAGNNNNNNNNNNNNNN_NN^.