Learn More
New England Biolabs, Inc. BpmI – 500 units
SDP
Catalog No. 50812252
Encompass
A restriction endonuclease that recognizes the sequence CTGGAGNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-252 Supplier New England Biolabs, Inc. Supplier No. R0565L
A restriction endonuclease that recognizes the sequence CTGGAGNNNNNNNNNNNNNN_NN^.