Learn More
Medchemexpress LLC BPN-15606 (free base) | 1914989-49-3 | MFCD32215307 | >98.0% | 418.48 g/mol | C23H23FN6O | 50 MG
SDP
Catalog No. 5000227722
Encompass_Preferred
BPN-15606 is a small-molecule γ-secretase modulator (CAS 1914989-49-3) used in research to reduce production of amyloid β peptides Aβ42 and Aβ40. It has demonstrated high potency in cell-based assays and oral activity in preclinical studies, and is supplied as a small research pack for in vitro and in vivo use.
  • Potent γ-secretase modulation with reported IC50 ≈ 7 nM (Aβ42) and 17 nM (Aβ40).
  • Orally active in preclinical animal models.
  • High purity suitable for research use; Certificate of Analysis available.
  • Suitable for in vitro cell assays and in vivo preclinical studies.
  • Provided in a compact research pack for convenient dosing and formulation.
Catalog No. 50-002-27722 Supplier Medchemexpress LLC Supplier No. HY11748250MG
BPN-15606 is a small-molecule γ-secretase modulator (CAS 1914989-49-3) used in research to reduce production of amyloid β peptides Aβ42 and Aβ40. It has demonstrated high potency in cell-based assays and oral activity in preclinical studies, and is supplied as a small research pack for in vitro and in vivo use.
  • Potent γ-secretase modulation with reported IC50 ≈ 7 nM (Aβ42) and 17 nM (Aβ40).
  • Orally active in preclinical animal models.
  • High purity suitable for research use; Certificate of Analysis available.
  • Suitable for in vitro cell assays and in vivo preclinical studies.
  • Provided in a compact research pack for convenient dosing and formulation.