Learn More
Medchemexpress LLC HY-117571 5mg Medchemexpress, BPN14770 CAS:1606974-33-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1175715MG
Encompass_Preferred
Medchemexpress, HY-117571 5mg BPN14770 CAS:1606974-33-7 BPN14770 is a selective phosphodiesterase 4D (PDE4D) allosteric inhibitor with IC50s of 7.8 nM and 7.4 nM for PDE4D7 and PDE4D3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9481