Learn More
Aviva Systems Biology Corp BPNT1 Antibody - N-terminal region (ARP58597_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58597_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BPNT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BPNT1. Alternate names: PIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62366